Assay Details
Target Gene Details
Entrez Gene ID: | 56550 |
Gene Name: | ubiquitin-conjugating enzyme E2D 2A |
Gene Aliases: |
1500034D03Rik, Ubc2e, Ube2d2, ubc4 |
Location: |
Chr.18:35771559-35807172 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2d2a | NM_019912.2 | NP_064296.1 | ||
| AK005355.1 | ||||
| AK136197.1 | BAE22869.1 | |||
| AK139623.1 | BAE24085.1 | |||
| AK146659.1 | ||||
| AK147568.1 | ||||
| AK148089.1 | BAE28338.1 | |||
| AK160567.1 | BAE35877.1 | |||
| AK162093.1 | BAE36719.1 | |||
| AK172088.1 | ||||
| AK190235.1 | ||||
| AK196425.1 | ||||
| AK198040.1 | ||||
| AK199554.1 | ||||
| AK204863.1 | ||||
| AK219213.1 | ||||
| BC003923.1 | AAH03923.1 | |||
| U62483.1 | AAB05772.1 |
More Information
Set Membership: |
|

Genomic Map