Assay Details
Target Gene Details
Entrez Gene ID: | 14119 |
Gene Name: | fibrillin 2 |
Gene Aliases: |
BC063774, Fib-2, mKIAA4226, sne, sy |
Location: |
Chr.18:58008623-58209926 on Build GRCm38 |
Assay Gene Location: | Within Exon 50 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbn2 | NM_010181.2 | 50 | 6470 | NP_034311.2 |
| AK220196.1 | 36 | 4305 | BAD90381.1 | |
| U20217.1 | 21 | 2521 | AAA62177.1 |
More Information
Set Membership: |
|

Genomic Map