Assay Details
Target Gene Details
Entrez Gene ID: | 14119 |
Gene Name: | fibrillin 2 |
Gene Aliases: |
BC063774, Fib-2, mKIAA4226, sne, sy |
Location: |
Chr.18:58008623-58209926 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 62 - Exon 62 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbn2 | NM_010181.2 | NP_034311.2 | ||
| AK220196.1 | BAD90381.1 |
More Information
Set Membership: |
|

Genomic Map