Assay Details
Target Gene Details
Entrez Gene ID: | 15531 |
Gene Name: | N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 1 |
Gene Aliases: |
1200015G06Rik, Hsst, NDST-1, b2b2230Clo |
Location: |
Chr.18:60684494-60748892 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndst1 | NM_008306.4 | NP_032332.2 | ||
| AF049894.1 | AAC17228.1 | |||
| AF074926.1 | AAD15980.1 | |||
| AK035642.1 | BAC29136.1 | |||
| AK147282.1 | BAE27818.1 | |||
| AK170041.1 | BAE41527.1 | |||
| AK171013.1 | BAE42184.1 | |||
| BC066098.1 | AAH66098.1 | |||
| BC079561.1 | AAH79561.1 |
More Information
Set Membership: |
|

Genomic Map