Assay Details
Target Gene Details
Entrez Gene ID: | 13824 |
Gene Name: | erythrocyte protein band 4.1 like 4a |
Gene Aliases: |
Epb4.1l4, Epb41l4a, NBL4 |
Location: |
Chr.18:33796327-34007678 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epb4.1l4a | NM_013512.2 | NP_038540.2 | ||
| AK140485.1 | BAE24405.1 | |||
| BC007166.1 | AAH07166.1 | |||
| BC013557.1 | AAH13557.1 | |||
| D28818.1 | BAA05978.1 |

Genomic Map