Assay Details
Target Gene Details
Entrez Gene ID: | 77619 |
Gene Name: | PRELI domain containing 2 |
Gene Aliases: |
1700003A01Rik, C330008K14Rik |
Location: |
Chr.18:41875696-41951194 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prelid2 | NM_029942.1 | NP_084218.1 | ||
| AK021193.1 | BAB32324.1 | |||
| BC119251.1 | ||||
| BC120711.1 | ||||
| BC145035.1 |

Genomic Map