Assay Details
Target Gene Details
Entrez Gene ID: | 19877 |
Gene Name: | Rho-associated coiled-coil containing protein kinase 1 |
Gene Aliases: |
1110055K06Rik, Rock-I |
Location: |
Chr.18:10064401-10181792 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rock1 | NM_009071.2 | 22 | 3083 | NP_033097.1 |
| AK085974.1 | 9 | 1197 | BAC39581.1 | |
| AK171291.1 | 2 | 108 | BAE42373.1 | |
| U58512.1 | 22 | 2608 | AAC53132.1 |
More Information
Set Membership: |
|

Genomic Map