Assay Details
Target Gene Details
Entrez Gene ID: | 74322 |
Gene Name: | CXXC finger 1 (PHD domain) |
Gene Aliases: |
2410002I16Rik, 5830420C16Rik, AI426635, Cfp1, Cgbp, PHF18 |
Location: |
Chr.18:74216093-74221496 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cxxc1 | NM_028868.3 | 4 | 568 | NP_083144.1 |
| AK010337.1 | 4 | 569 | BAB26862.1 | |
| AK083655.1 | 4 | 573 | BAC38986.1 | |
| AK146688.1 | 4 | 652 | BAE27361.1 | |
| AY099096.1 | 4 | 540 | AAM28246.1 | |
| BC030938.1 | 4 | 400 | AAH30938.1 |

Genomic Map