Assay Details
Target Gene Details
Entrez Gene ID: | 75964 |
Gene Name: | trafficking protein particle complex 8 |
Gene Aliases: |
5033403J15Rik, AI845423, D030074E01Rik, Trs85, mKIAA1012 |
Location: |
Chr.18:20817223-20896386 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc8 | NM_029491.2 | 1 | 124 | NP_083767.2 |
| NM_177038.3 | 1 | 124 | NP_796012.2 | |
| AK044854.1 | 1 | 409 | BAC32120.1 | |
| AK083747.1 | 1 | 124 | BAC39011.1 |

Genomic Map