Assay Details
Target Gene Details
Entrez Gene ID: | 19877 |
Gene Name: | Rho-associated coiled-coil containing protein kinase 1 |
Gene Aliases: |
1110055K06Rik, Rock-I |
Location: |
Chr.18:10064401-10181792 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rock1 | NM_009071.2 | 27 | 3745 | NP_033097.1 |
| AK050269.1 | 3 | 326 | BAC34154.1 | |
| AK171291.1 | 7 | 770 | BAE42373.1 | |
| U58512.1 | 27 | 3270 | AAC53132.1 |
More Information
Set Membership: |
|

Genomic Map