Assay Details
Target Gene Details
Entrez Gene ID: | 319262 |
Gene Name: | FCH and double SH3 domains 1 |
Gene Aliases: |
A030002D08Rik |
Location: |
Chr.18:37957434-37969814 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fchsd1 | NM_175684.4 | NP_783615.2 | ||
| AK079461.1 | BAC37653.1 | |||
| AK154732.1 | ||||
| BC057367.1 | AAH57367.1 |

Genomic Map