Assay Details
Target Gene Details
Entrez Gene ID: | 76217 |
Gene Name: | janus kinase and microtubule interacting protein 2 |
Gene Aliases: |
6430702L21Rik, AI850334, D930046L20Rik |
Location: |
Chr.18:43527096-43687773 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jakmip2 | NM_001163637.1 | NP_001157109.1 | ||
| AK133750.1 |

Genomic Map