Assay Details
Target Gene Details
Entrez Gene ID: | 319195 |
Gene Name: | ribosomal protein L17 |
Gene Aliases: |
- |
Location: |
Chr.18:74998978-75003381 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl17 | NM_001002239.3 | NP_001002239.2 | ||
| AA407762.1 | ||||
| AK002770.1 | BAB22345.1 | |||
| AK011132.1 | BAB27423.1 | |||
| AK011133.1 | BAB27424.1 | |||
| AK088443.1 | ||||
| AK137077.1 | ||||
| BC003896.1 | AAH03896.2 | |||
| BC052940.1 | AAH52940.1 | |||
| BC054424.1 | AAH54424.1 | |||
| BC090990.1 | AAH90990.1 | |||
| BC091759.1 | AAH91759.1 | |||
| BC092091.1 | AAH92091.1 | |||
| BC094640.1 | AAH94640.1 | |||
| BC106165.1 | AAI06166.1 |
More Information
Set Membership: |
|

Genomic Map