Assay Details
Target Gene Details
Entrez Gene ID: | 70361 |
Gene Name: | lectin, mannose-binding, 1 |
Gene Aliases: |
2610020P13Rik, AI326273, AU043785, C730041J05, ERGIC53, F5F8D, MCFD1, MR60, P58, gp58 |
Location: |
Chr.18:65980739-66002635 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lman1 | NM_001172062.1 | 12 | 1488 | NP_001165533.1 |
| NM_027400.3 | 12 | 1488 | NP_081676.1 | |
| AK011495.1 | 12 | 1474 | BAB27655.1 | |
| AK151599.1 | 12 | 1475 | BAE30539.1 | |
| AK151949.1 | 12 | 1475 | BAE30823.1 | |
| AK153554.1 | 12 | 1475 | BAE32091.1 | |
| BC057165.1 | 12 | 1497 | AAH57165.1 |
More Information
Set Membership: |
|

Genomic Map