Assay Details
Target Gene Details
Entrez Gene ID: | 24068 |
Gene Name: | steroid receptor RNA activator 1 |
Gene Aliases: |
AA959952, Sra, Srap, Straa1, Strra1 |
Location: |
Chr.18:36667187-36670311 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sra1 | NM_001164406.1 | NP_001157878.1 | ||
| NM_025291.3 | NP_079567.2 | |||
| NR_028355.1 | 1 | 209 | ||
| AF092039.1 | ||||
| AK007302.1 | BAB24943.1 | |||
| AK010375.1 | BAB26893.1 | |||
| AK136702.1 | ||||
| AK154232.1 | BAE32451.1 | |||
| BC048362.1 | AAH48362.1 | |||
| BX516162.1 | 1 | 158 | ||
| BY097874.1 |

Genomic Map