Assay Details
Target Gene Details
Entrez Gene ID: | 225358 |
Gene Name: | family with sequence similarity 13, member B |
Gene Aliases: |
2610024E20Rik, AW060714, AW546153 |
Location: |
Chr.18:34440571-34523046 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam13b | NM_146084.1 | 17 | 2543 | NP_666196.1 |
| AK029766.1 | 18 | 2659 | ||
| BC029030.1 | 1 | 191 | AAH29030.1 | |
| BC031465.1 | 17 | 2543 | AAH31465.1 |

Genomic Map