Assay Details
Target Gene Details
Entrez Gene ID: | 319211 |
Gene Name: | nucleolar protein 4 |
Gene Aliases: |
1700013J13Rik, 4930568N03Rik, Gm1262 |
Location: |
Chr.18:22693152-23041675 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol4 | NM_001161483.1 | NP_001154955.1 | ||
| NM_199024.2 | NP_950189.2 | |||
| AK029800.1 | BAC26621.1 | |||
| AK031008.1 | ||||
| AK031753.1 | 1 | 136 | ||
| AK086574.1 | BAC39694.1 | |||
| BC056377.1 | AAH56377.1 |

Genomic Map