Assay Details
Target Gene Details
Entrez Gene ID: | 225579 |
Gene Name: | solute carrier family 27 (fatty acid transporter), member 6 |
Gene Aliases: |
4732438L20Rik, FACVL2, FATP6, VLCS-H1 |
Location: |
Chr.18:58556240-58612869 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc27a6 | NM_001081072.1 | 8 | 1729 | NP_001074541.1 |
| AK028699.1 | 8 | 1731 | ||
| BC115435.1 | 8 | 1712 |
More Information
Set Membership: |
|

Genomic Map