Assay Details
Target Gene Details
Entrez Gene ID: | 14423 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.18:24205344-24286816 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt1 | NM_001160404.1 | NP_001153876.1 | ||
| NM_013814.3 | NP_038842.3 | |||
| AK083391.1 | ||||
| BC056215.1 | AAH56215.1 | |||
| BC090962.1 | AAH90962.1 | |||
| BY711053.1 | ||||
| U73820.1 | AAB58477.1 |
More Information
Set Membership: |
|

Genomic Map