Assay Details
Target Gene Details
Entrez Gene ID: | 225467 |
Gene Name: | protein geranylgeranyltransferase type I, beta subunit |
Gene Aliases: |
2010207C17Rik, 2610100E13, AI451237, AI551093, BGG1, GGT1 |
Location: |
Chr.18:46235474-46281089 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pggt1b | NM_172627.3 | NP_766215.1 | ||
| AK028188.1 | ||||
| AK081731.1 | BAC38314.1 | |||
| BC141021.1 |
More Information
Set Membership: |
|

Genomic Map