Assay Details
Target Gene Details
Entrez Gene ID: | 106957 |
Gene Name: | solute carrier family 39 (metal ion transporter), member 6 |
Gene Aliases: |
Ermelin |
Location: |
Chr.18:24579881-24603817 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc39a6 | NM_139143.3 | 9 | 2640 | NP_631882.2 |
| AB071697.1 | 8 | 2023 | BAB86300.1 | |
| AK028976.1 | 9 | 2599 | BAC26223.1 | |
| BC054780.1 | 8 | 1553 | ||
| BC055012.1 | 9 | 2310 | AAH55012.1 |
More Information
Set Membership: |
|

Genomic Map