Assay Details
Target Gene Details
Entrez Gene ID: | 240215 |
Gene Name: | solute carrier family 4, sodium bicarbonate cotransporter, member 9 |
Gene Aliases: |
AE4, D630003B07, D630024F24Rik |
Location: |
Chr.18:36528063-36556272 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc4a9 | NM_001271544.1 | NP_001258473.1 | ||
| NM_001271545.1 | NP_001258474.1 | |||
| NM_001271546.1 | NP_001258475.1 | |||
| NM_001271547.1 | NP_001258476.1 | |||
| NM_172830.2 | NP_766418.1 | |||
| AK050187.1 | ||||
| AK085269.1 | BAC39407.1 | |||
| AK085424.1 | ||||
| AK128930.1 | ||||
| AK142545.1 | BAE25101.1 | |||
| BC113179.1 | ||||
| BC113785.1 |
More Information
Set Membership: |
|

Genomic Map