Assay Details
Target Gene Details
Entrez Gene ID: | 67665 |
Gene Name: | dynactin 4 |
Gene Aliases: |
1110001K06Rik, 4930547K17Rik, C130039E17Rik, p62 |
Location: |
Chr.18:60526159-60558762 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dctn4 | NM_026302.3 | NP_080578.1 | ||
| AK016059.1 | BAB30095.1 | |||
| AK034292.1 | BAC28661.1 | |||
| AK050896.1 | ||||
| AK141855.1 | ||||
| AK160813.1 | BAE36028.1 | |||
| AK163240.1 | BAE37250.1 | |||
| AK168484.1 | BAE40371.1 | |||
| BC006677.1 | AAH06677.1 | |||
| BC034725.1 | AAH34725.1 |
More Information
Set Membership: |
|

Genomic Map