Assay Details
Target Gene Details
Entrez Gene ID: | 100042150 |
Gene Name: | neuregulin 2 |
Gene Aliases: |
Don1, NTAK |
Location: |
Chr.18:36016758-36199047 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrg2 | NM_001167891.1 | NP_001161363.1 | ||
| AW476657.1 | ||||
| AY227025.1 | AAO72522.1 | |||
| AY227026.1 | AAO72523.1 |
More Information
Set Membership: |
|

Genomic Map