Assay Details
Target Gene Details
Entrez Gene ID: | 225182 |
Gene Name: | retinoblastoma binding protein 8 |
Gene Aliases: |
9930104E21Rik, CtIP, RBBP-8, RIM, SAE2 |
Location: |
Chr.18:11633276-11743207 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbbp8 | NM_001081223.2 | NP_001074692.1 | ||
| NM_001252495.1 | NP_001239424.1 | |||
| NR_045526.1 | ||||
| NR_045527.1 | ||||
| AK037049.1 | ||||
| AK041384.1 | ||||
| AK077614.1 | ||||
| BC019665.1 | AAH19665.1 | |||
| BC050849.1 | AAH50849.1 | |||
| CX567593.1 |
More Information
Set Membership: |
|

Genomic Map