Assay Details
Target Gene Details
Entrez Gene ID: | 14815 |
Gene Name: | nuclear receptor subfamily 3, group C, member 1 |
Gene Aliases: |
GR, Grl-1, Grl1 |
Location: |
Chr.18:39410545-39519416 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr3c1 | NM_008173.3 | NP_032199.3 | ||
| AK037733.1 | ||||
| AK155200.1 | BAE33113.1 | |||
| AK156323.1 | BAE33674.1 | |||
| BC129912.1 | ||||
| BC129913.1 | ||||
| DQ504162.1 | ||||
| HM236293.1 | ||||
| X04435.1 | CAA28031.1 | |||
| X13358.1 | CAA31738.1 | |||
| X13359.1 | CAA31739.1 |

Genomic Map