Assay Details
Target Gene Details
Entrez Gene ID: | 225655 |
Gene Name: | slowmo homolog 1 (Drosophila) |
Gene Aliases: |
A230074A19Rik, BC019561 |
Location: |
Chr.18:67464835-67480581 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slmo1 | NM_144867.2 | 2 | 188 | NP_659116.1 |
| AK038905.1 | 2 | 147 | ||
| AK044163.1 | 2 | 154 | ||
| AK143220.1 | 2 | 172 | BAE25314.1 | |
| BC019561.1 | 2 | 137 | AAH19561.1 | |
| BY741062.1 | 2 | 189 |

Genomic Map