Assay Details
Target Gene Details
Entrez Gene ID: | 13527 |
Gene Name: | dystrobrevin alpha |
Gene Aliases: |
2210407P21Rik, DTN-A, Dtn, Gm19389, a-DB-1, adbn |
Location: |
Chr.18:23310020-23659719 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtna | NM_001285807.1 | NP_001272736.1 | ||
| NM_001285808.1 | NP_001272737.1 | |||
| NM_001285810.1 | NP_001272739.1 | |||
| NM_001285811.1 | NP_001272740.1 | |||
| NM_001285813.1 | NP_001272742.1 | |||
| NM_001285817.1 | NP_001272746.1 | |||
| NM_207650.4 | NP_997533.1 | |||
| AF143543.1 | AAD33914.1 | |||
| AF143544.1 | AAD33915.1 | |||
| AK019477.1 | BAB31746.1 | |||
| AK131649.1 | BAE20740.1 | |||
| AK133036.1 | BAE21478.1 | |||
| AK147279.1 | BAE27816.1 | |||
| AK147359.1 | BAE27865.1 | |||
| BC040364.1 | AAH40364.1 | |||
| BC050765.1 | AAH50765.1 | |||
| X95226.1 | CAA64518.1 | |||
| X95227.1 | CAA64519.1 |
More Information
Set Membership: |
|

Genomic Map