Assay Details
Target Gene Details
Entrez Gene ID: | 13527 |
Gene Name: | dystrobrevin alpha |
Gene Aliases: |
2210407P21Rik, DTN-A, Dtn, Gm19389, a-DB-1, adbn |
Location: |
Chr.18:23310020-23659719 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtna | NM_001285807.1 | 20 | 2320 | NP_001272736.1 |
| NM_001285808.1 | 16 | 1805 | NP_001272737.1 | |
| NM_001285810.1 | 16 | 1793 | NP_001272739.1 | |
| NM_001285813.1 | 17 | 1950 | NP_001272742.1 | |
| NM_001285817.1 | 15 | 1784 | NP_001272746.1 | |
| NM_207650.4 | 16 | 1906 | NP_997533.1 | |
| AK019477.1 | 20 | 2240 | BAB31746.1 | |
| AK131649.1 | 15 | 1677 | BAE20740.1 | |
| AK147279.1 | 17 | 1861 | BAE27816.1 | |
| AK147359.1 | 16 | 1750 | BAE27865.1 | |
| AK180960.1 | ||||
| BC040364.1 | 16 | 1873 | AAH40364.1 | |
| BC050765.1 | 15 | 1701 | AAH50765.1 | |
| X95226.1 | 16 | 1627 | CAA64518.1 |
More Information
Set Membership: |
|

Genomic Map