Assay Details
Target Gene Details
Entrez Gene ID: | 225655 |
Gene Name: | slowmo homolog 1 (Drosophila) |
Gene Aliases: |
A230074A19Rik, BC019561 |
Location: |
Chr.18:67464835-67480581 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slmo1 | NM_144867.2 | NP_659116.1 | ||
| AK038905.1 | 4 | 502 | ||
| AK143220.1 | BAE25314.1 | |||
| BC019561.1 | AAH19561.1 | |||
| BY741062.1 |

Genomic Map