Assay Details
Target Gene Details
Entrez Gene ID: | 66306 |
Gene Name: | family with sequence similarity 53, member C |
Gene Aliases: |
2810012G03Rik, Gm21179 |
Location: |
Chr.18:34758830-34773760 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam53c | NM_175104.4 | NP_780313.1 | ||
| AK044469.1 | BAC31938.1 | |||
| AK135712.1 | BAE22622.1 | |||
| AK154214.1 | BAE32439.1 | |||
| AK166841.1 | ||||
| AK219147.1 | 1 | 36 | ||
| BC044896.1 | AAH44896.1 | |||
| BC057111.1 | AAH57111.1 |

Genomic Map