Assay Details
Target Gene Details
Entrez Gene ID: | 106894 |
Gene Name: | HMG box domain containing 3 |
Gene Aliases: |
2510002C16Rik, A630042L21Rik, AI413166, mKIAA0194 |
Location: |
Chr.18:61131277-61177050 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmgxb3 | NM_134134.2 | 16 | 3063 | NP_598895.2 |
| NM_178277.1 | 15 | 3158 | NP_840061.1 | |
| AB045322.1 | 16 | 3064 | BAA97982.1 | |
| AK129085.1 | 12 | 2066 | BAC97895.1 | |
| AK132529.1 | 9 | 1542 | BAE21220.1 | |
| AK144993.1 | 15 | 2911 | ||
| BC025148.1 | AAH25148.1 | |||
| BC058089.1 | 13 | 2456 | AAH58089.1 | |
| BC060046.1 | 9 | 1488 | AAH60046.1 | |
| BC066029.1 | 10 | 2294 | AAH66029.1 | |
| BC079580.1 | 15 | 3106 | AAH79580.1 | |
| BG061435.2 | 1 | 154 |

Genomic Map