Assay Details
Target Gene Details
Entrez Gene ID: | 106894 |
Gene Name: | HMG box domain containing 3 |
Gene Aliases: |
2510002C16Rik, A630042L21Rik, AI413166, mKIAA0194 |
Location: |
Chr.18:61131277-61177050 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmgxb3 | NM_134134.2 | 17 | 3214 | NP_598895.2 |
| NM_178277.1 | 16 | 3309 | NP_840061.1 | |
| AB045322.1 | 17 | 3215 | BAA97982.1 | |
| AK129085.1 | 13 | 2217 | BAC97895.1 | |
| AK132529.1 | 10 | 1693 | BAE21220.1 | |
| AK144993.1 | ||||
| AK178113.1 | 1 | 73 | ||
| BC025148.1 | 2 | 177 | AAH25148.1 | |
| BC058089.1 | 14 | 2607 | AAH58089.1 | |
| BC060046.1 | 10 | 1639 | AAH60046.1 | |
| BC066029.1 | 11 | 2445 | AAH66029.1 | |
| BC079580.1 | 16 | 3257 | AAH79580.1 | |
| BG061435.2 | 2 | 305 |

Genomic Map