Assay Details
Target Gene Details
Entrez Gene ID: | 106894 |
Gene Name: | HMG box domain containing 3 |
Gene Aliases: |
2510002C16Rik, A630042L21Rik, AI413166, mKIAA0194 |
Location: |
Chr.18:61131277-61177050 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmgxb3 | NM_134134.2 | NP_598895.2 | ||
| NM_178277.1 | NP_840061.1 | |||
| AB045322.1 | BAA97982.1 | |||
| AK129085.1 | BAC97895.1 | |||
| AK132529.1 | BAE21220.1 | |||
| AK144993.1 | ||||
| AK178113.1 | ||||
| BC025148.1 | AAH25148.1 | |||
| BC058089.1 | AAH58089.1 | |||
| BC060046.1 | AAH60046.1 | |||
| BC066029.1 | AAH66029.1 | |||
| BC079580.1 | AAH79580.1 |

Genomic Map