Assay Details
Target Gene Details
Entrez Gene ID: | 106894 |
Gene Name: | HMG box domain containing 3 |
Gene Aliases: |
2510002C16Rik, A630042L21Rik, AI413166, mKIAA0194 |
Location: |
Chr.18:61131277-61177050 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmgxb3 | NM_134134.2 | NP_598895.2 | ||
| NM_178277.1 | NP_840061.1 | |||
| AB045322.1 | BAA97982.1 | |||
| AK034086.1 | BAC28577.1 | |||
| AK041866.1 | BAC31085.1 | |||
| AK144993.1 | ||||
| BC079580.1 | AAH79580.1 |

Genomic Map