Assay Details
Target Gene Details
Entrez Gene ID: | 106894 |
Gene Name: | HMG box domain containing 3 |
Gene Aliases: |
2510002C16Rik, A630042L21Rik, AI413166, mKIAA0194 |
Location: |
Chr.18:61131277-61177050 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmgxb3 | NM_134134.2 | 6 | 1450 | NP_598895.2 |
| NM_178277.1 | 6 | 1449 | NP_840061.1 | |
| AB045322.1 | 6 | 1450 | BAA97982.1 | |
| AK041866.1 | 6 | 1428 | BAC31085.1 | |
| AK129085.1 | 3 | 357 | BAC97895.1 | |
| AK144993.1 | 6 | 1434 | ||
| BC058089.1 | 4 | 747 | AAH58089.1 | |
| BC079580.1 | 6 | 1397 | AAH79580.1 | |
| BU611563.1 | 3 | 561 |

Genomic Map