Assay Details
Target Gene Details
Entrez Gene ID: | 225266 |
Gene Name: | kelch-like 14 |
Gene Aliases: |
6330403N15Rik, AI785250, printor |
Location: |
Chr.18:21550377-21672932 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhl14 | NM_001081403.1 | NP_001074872.1 | ||
| AK018108.1 | BAB31073.1 | |||
| AK173163.1 | BAD32441.1 |

Genomic Map