Assay Details
Target Gene Details
Entrez Gene ID: | 225392 |
Gene Name: | RELT-like 2 |
Gene Aliases: |
4631403P03Rik, ependolin |
Location: |
Chr.18:37955084-37959179 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rell2 | NM_153793.2 | NP_722488.2 | ||
| AK044097.1 | BAC31774.1 | |||
| BC038500.1 | AAH38500.1 |

Genomic Map