Assay Details
Target Gene Details
Entrez Gene ID: | 22139 |
Gene Name: | transthyretin |
Gene Aliases: |
AA408768, AI787086, D17860, prealbumin |
Location: |
Chr.18:20665250-20674326 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttr | NM_013697.5 | NP_038725.1 | ||
| AK014454.1 | BAB29362.1 | |||
| AK018701.1 | BAB31352.1 | |||
| AK050155.1 | BAC34098.1 | |||
| AK075588.1 | BAC35839.1 | |||
| BC024702.1 | AAH24702.1 | |||
| BC086926.1 | AAH86926.1 | |||
| CK627038.1 | ||||
| D89076.1 | BAA13757.1 | |||
| X03351.1 | CAA27057.1 |
More Information
Set Membership: |
|

Genomic Map