Assay Details
Target Gene Details
Entrez Gene ID: | 67951 |
Gene Name: | tubulin, beta 6 class V |
Gene Aliases: |
2310057H16Rik, BB220206 |
Location: |
Chr.18:67390731-67402749 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb6 | NM_026473.2 | NP_080749.2 | ||
| AK009958.1 | ||||
| AK144815.1 | BAE26077.1 | |||
| AK151643.1 | BAE30573.1 | |||
| BC008225.1 | AAH08225.1 |
More Information
Set Membership: |
|

Genomic Map