Assay Details
Target Gene Details
Entrez Gene ID: | 76594 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 18 |
Gene Aliases: |
2700075B01Rik, AU041129 |
Location: |
Chr.18:35671103-35708421 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc18 | NM_029669.4 | 4 | 596 | NP_083945.1 |
| AK012521.1 | 4 | 549 | BAB28293.1 | |
| AK030483.1 | 4 | 574 | BAC26985.1 | |
| BC026452.1 | 4 | 558 | AAH26452.1 |

Genomic Map