Assay Details
Target Gene Details
Entrez Gene ID: | 170826 |
Gene Name: | peroxisome proliferative activated receptor, gamma, coactivator 1 beta |
Gene Aliases: |
4631412G21Rik, Perc |
Location: |
Chr.18:61291445-61401462 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppargc1b | NM_133249.2 | NP_573512.1 | ||
| AF453324.1 | AAL47054.1 | |||
| AK028464.1 | BAC25964.1 | |||
| AK086111.1 | ||||
| BC150699.1 |
More Information
Set Membership: |
|

Genomic Map