Assay Details
Target Gene Details
Entrez Gene ID: | 19762 |
Gene Name: | Ras-like without CAAX 2 |
Gene Aliases: |
RIBA, Rin, Roc2 |
Location: |
Chr.18:30974314-31317128 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rit2 | NM_009065.2 | 4 | 559 | NP_033091.1 |
| AF084463.1 | 4 | 458 | AAD13022.1 | |
| AK040743.1 | 4 | 518 | BAC30690.1 | |
| AK046425.1 | 4 | 419 | BAC32724.1 | |
| AK046484.1 | 4 | 341 | BAC32750.1 | |
| AK138206.1 | 4 | 370 | BAE23581.1 | |
| AK139394.1 | 4 | 456 | BAE23992.1 | |
| AK140726.1 | 4 | 565 | BAE24457.1 | |
| AK162862.1 | 4 | 462 | BAE37088.1 | |
| AK162953.1 | 4 | 515 | BAE37131.1 | |
| BC018267.1 | 4 | 559 | AAH18267.1 | |
| U71202.1 | 4 | 517 | AAB42212.1 |
More Information
Set Membership: |
|

Genomic Map