Assay Details
Target Gene Details
Entrez Gene ID: | 67788 |
Gene Name: | SWI5 dependent recombination repair 1 |
Gene Aliases: |
6330577E15Rik, Meir5 |
Location: |
Chr.19:47731733-47735588 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sfr1 | NM_026377.2 | NP_080653.2 | ||
| AB597963.1 | ||||
| AK004073.1 | BAB23154.1 | |||
| AK011531.1 | BAB27680.1 | |||
| AK014266.1 | BAB29232.1 | |||
| AK016079.1 | BAB30105.1 | |||
| AK077875.1 | BAC37045.1 | |||
| AK078147.1 | BAC37148.1 | |||
| AK146810.1 | BAE27450.1 | |||
| AK167406.1 | BAE39495.1 | |||
| AK168068.1 | BAE40043.1 | |||
| AK168427.1 | BAE40336.1 | |||
| AK217479.1 | ||||
| BC024403.1 | AAH24403.1 |
More Information
Set Membership: |
|

Genomic Map