Assay Details
Target Gene Details
Entrez Gene ID: | 56327 |
Gene Name: | ADP-ribosylation factor-like 2 |
Gene Aliases: |
2610009M23Rik, AI115441, AW553335 |
Location: |
Chr.19:6134389-6141137 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl2 | NM_019722.3 | NP_062696.2 | ||
| AF143680.1 | AAD33908.1 | |||
| AK011366.1 | BAB27572.1 | |||
| AK181583.1 | ||||
| BC060259.1 | AAH60259.1 |

Genomic Map