Assay Details
Target Gene Details
Entrez Gene ID: | 226090 |
Gene Name: | endoplasmic reticulum metallopeptidase 1 |
Gene Aliases: |
D19Ertd410e, D19Wsu12e, Fxna, mKIAA1815 |
Location: |
Chr.19:29609883-29648484 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ermp1 | NM_001081213.1 | 9 | 1638 | NP_001074682.1 |
| AK050284.1 | 9 | 1634 | ||
| AK081674.1 | 8 | 1155 | BAC38286.1 | |
| AK129447.1 | 9 | 1566 | BAC98257.1 | |
| AK137206.1 | 9 | 1594 | BAE23269.1 | |
| BC151181.1 | 9 | 1597 | ||
| BC151182.1 | 9 | 1597 | ||
| BC157914.1 | 9 | 1597 |
More Information
Set Membership: |
|

Genomic Map