Assay Details
Target Gene Details
Entrez Gene ID: | 332396 |
Gene Name: | potassium channel, subfamily K, member 18 |
Gene Aliases: |
Gm781, Tresk, Tresk-2, Trik |
Location: |
Chr.19:59219648-59237370 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk18 | NM_207261.3 | NP_997144.1 | ||
| AK139600.1 | BAE24080.1 | |||
| AK162136.1 | BAE36746.1 | |||
| AY325301.1 | AAQ91836.1 | |||
| AY542902.1 | AAS48426.1 | |||
| BC104132.1 | AAI04133.1 | |||
| BC104133.1 | AAI04134.1 | |||
| BC115705.1 | ||||
| BC115887.1 | ||||
| BC127136.1 | ||||
| BC127137.1 |
More Information
Set Membership: |
|

Genomic Map