Assay Details
Target Gene Details
Entrez Gene ID: | 16530 |
Gene Name: | potassium channel, subfamily K, member 7 |
Gene Aliases: |
2310014G03Rik, Kcnk6, Kcnk8, Knot, Mlk3 |
Location: |
Chr.19:5704476-5707405 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk7 | NM_010609.2 | NP_034739.2 | ||
| AB015729.1 | BAA35074.1 | |||
| AF012324.1 | AAF21603.1 | |||
| AF022820.1 | AAD09337.1 | |||
| AF110521.1 | AAD29577.1 | |||
| AK160180.1 | ||||
| BC106993.2 | AAI06994.1 |
More Information
Set Membership: |
|

Genomic Map