Assay Details
Target Gene Details
Entrez Gene ID: | 57344 |
Gene Name: | arsenic (+3 oxidation state) methyltransferase |
Gene Aliases: |
2310045H08Rik, Cyt19 |
Location: |
Chr.19:46707359-46741099 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| As3mt | NM_020577.2 | 4 | 335 | NP_065602.2 |
| AF166383.1 | 4 | 305 | AAF00618.1 | |
| AK002801.1 | 4 | 268 | ||
| AK006571.1 | 4 | 320 | ||
| AK009814.1 | 3 | 322 | ||
| AK011320.1 | 4 | 287 | ||
| BC145663.1 | 4 | 277 | ||
| BY287843.1 | 4 | 335 |
More Information
Set Membership: |
|

Genomic Map