Assay Details
Target Gene Details
Entrez Gene ID: | 67674 |
Gene Name: | tRNA methyltransferase 11-2 |
Gene Aliases: |
0610038D11Rik, Trm112p |
Location: |
Chr.19:6909698-6911028 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt112 | NM_001166370.1 | 1 | 79 | NP_001159842.1 |
| NM_026306.3 | 1 | 79 | NP_080582.3 | |
| BC016191.1 | AAH16191.1 | |||
| BC087959.1 | 1 | 79 | AAH87959.1 | |
| DV654053.1 |

Genomic Map